Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204100.1 | NP_989431.1 | 2 - 3 | 225 | 85 | |
XM_015287950.2 | XP_015143436.1 | 1 - 2 | 207 | 85 | ||
GenBank mRNA | AB021166.1 | - | 1 - 2 | 210 | 85 | |
AB059657.1 | - | 2 - 3 | 225 | 85 | ||
AJ719978.1 | - | 2 - 3 | 239 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |