Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204132.1 | NP_989463.1 | 1 - 2 | 222 | 75 | |
GenBank mRNA | AY037891.1 | - | 1 - 2 | 222 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |