Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204160.1 | NP_989491.1 | 4 - 5 | 1470 | 143 | |
GenBank mRNA | AB023291.1 | - | 4 - 5 | 1411 | 143 | |
D88349.1 | - | 4 - 5 | 1470 | 143 | ||
MG672516.1 | - | 4 - 5 | 1364 | 143 | ||
S81675.1 | - | 4 - 5 | 1406 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |