Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204162.1 | NP_989493.1 | 3 - 4 | 846 | 91 | |
GenBank mRNA | AF092538.1 | - | 3 - 4 | 846 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |