Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204177.2 | NP_989508.1 | 6 - 7 | 898 | 80 | |
GenBank mRNA | AF069771.1 | - | 6 - 7 | 763 | 80 | |
AF218799.1 | - | 6 - 7 | 899 | 80 | ||
BX929571.2 | - | 6 - 7 | 800 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |