Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204179.1 | NP_989510.1 | 4 - 5 | 490 | 60 | |
XM_025152069.1 | XP_025007837.1 | 3 - 4 | 6393 | 60 | ||
GenBank mRNA | U28145.1 | - | 4 - 5 | 490 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |