Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204180.1 | NP_989511.1 | 4 - 5 | 468 | 71 | |
GenBank mRNA | M11619.1 | - | 4 - 5 | 468 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |