Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FECH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204196.1 | NP_989527.1 | 3 - 4 | 254 | 64 | |
GenBank mRNA | U68033.1 | - | 3 - 4 | 254 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |