Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204204.1 | NP_989535.1 | 2 - 3 | 1079 | 112 | |
GenBank mRNA | AF129809.1 | - | 2 - 3 | 1079 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |