Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204244.1 | NP_989575.1 | 8 - 9 | 1320 | 86 | |
XM_015286915.2 | XP_015142401.1 | 8 - 9 | 1361 | 86 | ||
GenBank mRNA | AY034433.1 | - | 8 - 9 | 1320 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |