Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204248.2 | NP_989579.2 | 3 - 4 | 866 | 83 | |
GenBank mRNA | AF165889.1 | - | 3 - 4 | 1062 | 83 | |
BU327747.1 | - | 3 - 4 | 297 | 83 | ||
FJ417094.1 | - | 3 - 4 | 882 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |