Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204249.1 | NP_989580.1 | 724 | 137 | ||
GenBank mRNA | L24551.1 | - | 724 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus |