Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204283.1 | NP_989614.1 | 1 - 2 | 112 | 110 | |
GenBank mRNA | AF008194.1 | - | 1 - 2 | 112 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |