Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204301.1 | NP_989632.1 | 7 - 8 | 638 | 97 | |
GenBank mRNA | AF228284.1 | - | 7 - 8 | 638 | 97 | |
AF407671.1 | - | 8 - 9 | 654 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |