Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204336.2 | NP_989667.2 | 705 | 70 | ||
GenBank mRNA | AF182403.1 | - | 70 | |||
AF346628.1 | - | 70 | ||||
BX935899.1 | - | 706 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus |