Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204347.1 | NP_989678.1 | 5 - 6 | 563 | 71 | |
GenBank mRNA | AF351203.1 | - | 5 - 6 | 563 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |