Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204354.1 | NP_989685.1 | 2 - 3 | 991 | 58 | |
GenBank mRNA | AY207465.1 | - | 1 - 2 | 140 | 58 | |
AY278202.1 | - | 2 - 3 | 991 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |