Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204355.1 | NP_989686.1 | 4 - 5 | 363 | 80 | |
GenBank mRNA | AB050010.1 | - | 4 - 5 | 363 | 80 | |
BX934974.2 | - | 4 - 5 | 476 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |