Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204367.2 | NP_989698.2 | 3 - 4 | 707 | 72 | |
XM_015298410.2 | XP_015153896.1 | 3 - 4 | 1836 | 72 | ||
GenBank mRNA | AY139091.1 | - | 3 - 4 | 678 | 72 | |
BU323753.1 | - | 3 - 4 | 423 | 72 | ||
BX935023.2 | - | 3 - 4 | 538 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |