Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204382.1 | NP_989713.1 | 2 - 3 | 441 | 88 | |
XM_025152732.1 | XP_025008500.1 | 2 - 3 | 115 | 85 | ||
GenBank mRNA | AB095027.2 | - | 2 - 3 | 441 | 88 |
Set Membership: |
Amplicon length between 71 and 85 Amplicon length between 86 and 100 Gallus gallus Probe spans exons |