Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204382.1 | NP_989713.1 | 3 - 4 | 521 | 126 | |
XM_025152732.1 | XP_025008500.1 | 3 - 4 | 192 | 126 | ||
GenBank mRNA | AB095027.2 | - | 3 - 4 | 521 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |