Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204389.2 | NP_989720.3 | 5 - 6 | 892 | 74 | |
XM_025153787.1 | XP_025009555.1 | 5 - 6 | 1128 | 74 | ||
GenBank mRNA | D89097.2 | - | 5 - 6 | 892 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |