Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204391.1 | NP_989722.1 | 6 - 7 | 630 | 111 | |
XM_015283077.2 | XP_015138563.1 | 7 - 8 | 832 | 111 | ||
XM_015283079.2 | XP_015138565.1 | 7 - 8 | 817 | 111 | ||
XM_025147006.1 | XP_025002774.1 | 6 - 7 | 795 | 111 | ||
GenBank mRNA | AY278230.1 | - | 6 - 7 | 630 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |