Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204409.1 | NP_989740.1 | 2 - 3 | 382 | 76 | |
GenBank mRNA | AB105911.2 | - | 2 - 3 | 422 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |