Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204414.2 | NP_989745.2 | 3 - 4 | 440 | 113 | |
XM_015296809.1 | XP_015152295.1 | 3 - 4 | 435 | 113 | ||
XM_015296811.1 | XP_015152297.1 | 3 - 4 | 435 | 113 | ||
XM_025142477.1 | XP_024998245.1 | 3 - 4 | 409 | 113 | ||
GenBank mRNA | CN232738.1 | - | 3 - 4 | 440 | 113 | |
CR391085.1 | - | 3 - 4 | 395 | 113 | ||
D26328.1 | - | 3 - 4 | 395 | 113 | ||
L13032.1 | - | 3 - 4 | 389 | 113 | ||
L20500.1 | - | 3 - 4 | 247 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |