Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204414.2 | NP_989745.2 | 4 - 5 | 532 | 63 | |
XM_015296809.1 | XP_015152295.1 | 4 - 5 | 527 | 63 | ||
XM_015296811.1 | XP_015152297.1 | 4 - 5 | 527 | 63 | ||
XM_025142477.1 | XP_024998245.1 | 4 - 5 | 501 | 63 | ||
GenBank mRNA | CN232738.1 | - | 4 - 5 | 532 | 63 | |
CR391085.1 | - | 4 - 5 | 487 | 63 | ||
D26328.1 | - | 4 - 5 | 487 | 63 | ||
L13032.1 | - | 4 - 5 | 481 | 63 | ||
L20500.1 | - | 4 - 5 | 339 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |