Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204418.1 | NP_989749.1 | 6 - 7 | 856 | 142 | |
GenBank mRNA | JQ277718.1 | - | 2 - 3 | 142 | ||
L08047.1 | - | 6 - 7 | 856 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |