Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204428.1 | NP_989759.1 | 2 - 3 | 1398 | 98 | |
GenBank mRNA | AJ535752.1 | - | 2 - 3 | 1398 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |