Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204429.1 | NP_989760.1 | 932 | 99 | ||
GenBank mRNA | AB071038.1 | - | 932 | 99 | ||
BX935108.1 | - | 921 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus |