Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204441.1 | NP_989772.1 | 2 - 3 | 230 | 78 | |
XM_015276809.2 | XP_015132295.1 | 3 - 4 | 356 | 78 | ||
XM_015276810.2 | XP_015132296.1 | 3 - 4 | 4535 | 78 | ||
XM_025153303.1 | XP_025009071.1 | 3 - 4 | 346 | 78 | ||
XM_025153305.1 | XP_025009073.1 | 3 - 4 | 356 | 78 | ||
GenBank mRNA | AF465270.1 | - | 2 - 3 | 230 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |