Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204485.2 | NP_989816.2 | 8 - 9 | 923 | 103 | |
XM_015277227.2 | XP_015132713.1 | 8 - 9 | 835 | 103 | ||
GenBank mRNA | AB109635.1 | - | 7 - 8 | 768 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |