Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204507.1 | NP_989838.1 | 4 - 5 | 580 | 110 | |
GenBank mRNA | AF373778.1 | - | 4 - 5 | 580 | 110 | |
BX933105.1 | - | 4 - 5 | 459 | 110 | ||
BX936181.1 | - | 2 - 3 | 233 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |