Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204510.1 | NP_989841.1 | 1 - 2 | 101 | 86 | |
XM_015288348.2 | XP_015143834.1 | 1 - 2 | 129 | 86 | ||
XM_015288350.2 | XP_015143836.1 | 1 - 2 | 129 | 86 | ||
GenBank mRNA | AY429472.1 | - | 1 - 2 | 64 | 86 | |
AY429473.1 | - | 1 - 2 | 64 | 86 | ||
AY451855.1 | - | 1 - 2 | 101 | 86 | ||
BX930596.2 | - | 1 - 2 | 50 | 86 | ||
BX936268.1 | - | 1 - 2 | 130 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |