Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204525.1 | NP_989856.1 | 4 - 5 | 542 | 133 | |
XM_015299568.2 | XP_015155054.1 | 5 - 6 | 681 | 133 | ||
GenBank mRNA | Y07811.1 | - | 542 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |