Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFI1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205091.1 | NP_990422.1 | 2 - 3 | 190 | 70 | |
GenBank mRNA | Y10898.1 | - | 190 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |