Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205097.1 | NP_990428.1 | 3 - 4 | 957 | 83 | |
XM_015286981.2 | XP_015142467.1 | 4 - 5 | 6769 | 83 | ||
XM_015286983.2 | XP_015142469.1 | 4 - 5 | 6610 | 83 | ||
XM_015286984.2 | XP_015142470.1 | 3 - 4 | 6592 | 83 | ||
XM_015286985.2 | XP_015142471.1 | 3 - 4 | 6580 | 83 | ||
XM_015286986.2 | XP_015142472.1 | 2 - 3 | 6400 | 83 | ||
GenBank mRNA | AF000138.1 | - | 3 - 4 | 957 | 83 | |
CR386643.1 | - | 3 - 4 | 811 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |