Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205258.2 | NP_990589.1 | 5 - 6 | 556 | 103 | |
GenBank mRNA | CF257199.1 | - | 5 - 6 | 558 | 103 | |
X66906.1 | - | 453 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |