Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205265.1 | NP_990596.1 | 4 - 5 | 669 | 114 | |
GenBank mRNA | X17199.1 | - | 669 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |