Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005837.1 | NP_001005837.1 | 4 - 5 | 954 | 65 | |
XM_015279148.2 | XP_015134634.1 | 4 - 5 | 1062 | 65 | ||
GenBank mRNA | AJ720466.1 | - | 4 - 5 | 954 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |