Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006128.1 | NP_001006128.1 | 1 - 2 | 173 | 120 | |
GenBank mRNA | AJ719291.1 | - | 1 - 2 | 173 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |