Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHODH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006131.2 | NP_001006131.1 | 5 - 6 | 898 | 73 | |
GenBank mRNA | AJ721004.1 | - | 5 - 6 | 899 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |