Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006140.2 | NP_001006140.2 | 5 - 6 | 528 | 135 | |
GenBank mRNA | AJ720805.1 | - | 5 - 6 | 529 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |