Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006145.1 | NP_001006145.1 | 5 - 6 | 426 | 87 | |
GenBank mRNA | AJ721112.1 | - | 5 - 6 | 426 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |