Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPSAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006165.1 | NP_001006165.1 | 5 - 6 | 512 | 88 | |
XM_015294183.2 | XP_015149669.1 | 4 - 5 | 422 | 88 | ||
GenBank mRNA | AJ719908.1 | - | 5 - 6 | 512 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |