Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006172.2 | NP_001006172.1 | 4 - 5 | 877 | 83 | |
GenBank mRNA | AJ719408.1 | - | 4 - 5 | 879 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |