Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006174.1 | NP_001006174.1 | 1 - 2 | 322 | 92 | |
XM_015294232.2 | XP_015149718.1 | 1 - 2 | 399 | 92 | ||
GenBank mRNA | AJ719564.1 | - | 1 - 2 | 322 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |