Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006188.1 | NP_001006188.1 | 4 - 5 | 731 | 111 | |
XM_015275281.2 | XP_015130767.1 | 4 - 5 | 805 | 111 | ||
XM_025155454.1 | XP_025011222.1 | 4 - 5 | 710 | 111 | ||
GenBank mRNA | AJ719798.1 | - | 4 - 5 | 731 | 111 | |
BX933911.2 | - | 4 - 5 | 713 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |