Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006189.1 | NP_001006189.1 | 2 - 3 | 258 | 74 | |
GenBank mRNA | AJ720363.1 | - | 2 - 3 | 258 | 74 | |
BX935300.1 | - | 2 - 3 | 215 | 74 | ||
BX936274.1 | - | 2 - 3 | 215 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |