Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006201.2 | NP_001006201.1 | 1 - 2 | 126 | 81 | |
GenBank mRNA | AJ450765.1 | - | 1 - 2 | 127 | 81 | |
AJ720053.1 | - | 1 - 2 | 103 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |