Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006202.1 | NP_001006202.1 | 11 - 12 | 1520 | 80 | |
GenBank mRNA | AJ719725.1 | - | 11 - 12 | 1520 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |